Learn More
Hach Company Free Chlorine Chemkey Reagents
Catalog No. 501997337 Shop All Hach Company Products
Encompass
Free Chlorine Chemkeys, range 0.04 - 4.00 mg/L Cl2. For SL1000 Portable Parallel Analyzer (PPA). Box of 25.USEPA accepted for reporting in drinking water analysis using Hach Method 10260.
Catalog No. 50-199-7337 Supplier Hach Company Supplier No. 9429000
Free Chlorine Chemkeys, range 0.04 - 4.00 mg/L Cl2. For SL1000 Portable Parallel Analyzer (PPA). Box of 25.USEPA accepted for reporting in drinking water analysis using Hach Method 10260.