Learn More
Hach Company High Range Orthophosphate Chemkey Reagents (box of 25)
Supplier:  Hach Company 8636500
Encompass
High Range Orthophosphate Chemkeys, range 2.0 - 30.0 mg/L PO4. For SL1000 Portable Parallel Analyzer (PPA). Box of 25.
Catalog No. 50-199-7276