Learn More
Hach Company Low Range Orthophosphate Chemkey Reagents (box of 25)
Supplier:  Hach Company 8636600
Encompass
Low Range Orthophosphate Chemkeys, range 0.20 - 4.00 mg/L PO4. For SL1000 Portable Parallel Analyzer (PPA). Box of 25.
Catalog No. 50-199-7277