Learn More
Hach Company Low Range Total Alkalinity Chemkey Reagents (box of 25)
Supplier:  Hach Company 8636200
Encompass
Low Range Total Alkalinity Chemkeys, range 20 - 200 mg/L CaCO3. For SL1000 Portable Parallel Analyzer (PPA). Box of 25.
Catalog No. 50-199-7273