Learn More
Azenta US Inc ASSY PLATE 384WL BLKFR BLKWL FFrameStar 384 Well Skirted PCR Plate black PP wells black PC frame cut corner A24 50 plates per case
Supplier:  Azenta US Inc 4TI0386
Encompass
ASSY PLATE 384WL BLKFR BLKWL FFrameStar 384 Well Skirted PCR Plate black PP wells black PC frame cut corner A24 50 plates per case
Catalog No. NC3243056