Learn More
Azenta US Inc ASSY SHEET ADH OPT CL QPCR 140qPCR Adhesive Seal optically clear film pressure activated adhesive 100 sheets 140 x 77mm per case
Catalog No. NC3243062
Encompass
ASSY SHEET ADH OPT CL QPCR 140qPCR Adhesive Seal optically clear film pressure activated adhesive 100 sheets 140 x 77mm per case
Catalog No. NC3243062 Supplier Azenta US Inc Supplier No. 4TI0560
ASSY SHEET ADH OPT CL QPCR 140qPCR Adhesive Seal optically clear film pressure activated adhesive 100 sheets 140 x 77mm per case