Learn More
Hach Company Contacting Conductivity Sensor, Cell Constant K=10, 3/4 inch Kynar Compression Fitting
Supplier:  Hach Company 3422E3A
Encompass
Nominal Cell Constant (K) = 10, 3/4 inch NPT Kynar (PVDF) Compression Fitting, 6m (20ft) Analog Cable, Titanium Electrode
Catalog No. 50-199-7441