Learn More
Nova Biomedical Corporation Nova Biomedical 61371 Waste Tubing Harness, Efficient Waste Management, Twist-lock Connector, Each
Catalog No. NC2165688
Encompass
The Nova Biomedical Waste Tubing Harness is a crucial component for the BioProfile FLEX2 Automated Cell Culture Analyzer. It enhances laboratory safety by facilitating efficient waste management systems, eliminating direct handling of hazardous materials. Its design enables easy snap-in replacement and includes secure connectors, optimizing maintenance and operational efficiency.
This item is not returnable. View return policy
Catalog No. NC2165688 Supplier Nova Biomedical Corporation Supplier No. 61371
The Nova Biomedical Waste Tubing Harness is a crucial component for the BioProfile FLEX2 Automated Cell Culture Analyzer. It enhances laboratory safety by facilitating efficient waste management systems, eliminating direct handling of hazardous materials. Its design enables easy snap-in replacement and includes secure connectors, optimizing maintenance and operational efficiency.