Learn More
LECO Corporation Glass Tube, requires one 765-976 O-ring and one 772-520 O-ring for sealing
Catalog No. NC1054932
Encompass
Glass Tube, requires one 765-976 O-ring and one 772-520 O-ring for sealing.
Catalog No. NC1054932 Supplier LECO Corporation Supplier No. 619591740
Glass Tube, requires one 765-976 O-ring and one 772-520 O-ring for sealing.