Learn More
Waters Corp Waters Corporation Tubing Assembly, MP35N, 0.004 in. ID, 19 in.
Catalog No. NC1268734 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement part designed for use with H-Class Bio CM-A Systems. It is constructed from MP35N material and provided with low pressure fittings.
  • Constructed from MP35N material
  • Internal diameter of 0.004 in.
  • Length of 19 in.
  • Designed for UPLC systems
  • Suitable for low pressure applications
  • Replacement schedule as required
Catalog No. NC1268734 Supplier Waters Corp Supplier No. 700005483
This tubing assembly is a replacement part designed for use with H-Class Bio CM-A Systems. It is constructed from MP35N material and provided with low pressure fittings.
  • Constructed from MP35N material
  • Internal diameter of 0.004 in.
  • Length of 19 in.
  • Designed for UPLC systems
  • Suitable for low pressure applications
  • Replacement schedule as required