Learn More
Waters Corp Waters Tubing, Stainless Steel, 0.003 in. ID, 19 in. Length
Catalog No. NC1397055 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement part for high-pressure systems, specifically designed for I-Class CM-A Systems. It is constructed from stainless steel and comes with high-pressure fittings.
  • Constructed from stainless steel.
  • Features an internal diameter of 0.003 in.
  • Has a length of 19 in.
  • Suitable for UPLC system types.
  • Recommended replacement schedule is as required.
Catalog No. NC1397055 Supplier Waters Corp Supplier No. 700008868
This tubing assembly is a replacement part for high-pressure systems, specifically designed for I-Class CM-A Systems. It is constructed from stainless steel and comes with high-pressure fittings.
  • Constructed from stainless steel.
  • Features an internal diameter of 0.003 in.
  • Has a length of 19 in.
  • Suitable for UPLC system types.
  • Recommended replacement schedule is as required.