Learn More
Waters Corp Tubing Assembly, Stainless Steel, 0.004 in. ID, 14.5 in. Length
Catalog No. NC2457711 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for use in ACQUITY UPLC and H-Class Systems. It is made from stainless steel and features low-pressure fittings, with an internal diameter of 0.004 in. and a length of 14.5 in.
  • Provided with low pressure fittings.
  • Constructed from stainless steel.
  • Features an internal diameter of 0.004 in.
  • Measures 14.5 in. in length.
  • Suitable for ACQUITY UPLC and H-Class Systems.
  • Functions as a replacement tubing assembly.
Catalog No. NC2457711 Supplier Waters Corp Supplier No. 700005477
This tubing assembly is designed for use in ACQUITY UPLC and H-Class Systems. It is made from stainless steel and features low-pressure fittings, with an internal diameter of 0.004 in. and a length of 14.5 in.
  • Provided with low pressure fittings.
  • Constructed from stainless steel.
  • Features an internal diameter of 0.004 in.
  • Measures 14.5 in. in length.
  • Suitable for ACQUITY UPLC and H-Class Systems.
  • Functions as a replacement tubing assembly.