Learn More
Apexbio Technology LLC AST-1306 897383-62-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061491
Encompass_Preferred
AST-1306 (CAS 897383-62-9) is an inhibitor of epidermal growth factor receptor (EGFR) and HER2 receptor tyrosine kinases In biochemical assays AST-1306 inhibits EGFR and HER2 kinase activity with IC50 values of 0 5 nM and 3 nM respectively It also demonstrates inhibitory activity against EGFR mutations such as T790M/L858R reducing receptor phosphorylation and downstream signaling in both cell-free and cellular systems In various animal tumor models including ErbB2-overexpressing and SK-OV-3 xenografts AST-1306 suppresses tumor growth and signaling pathways supporting its use in EGFR- and HER2-related oncology research
Catalog No. 50-000-61491 Supplier Apexbio Technology LLC Supplier No. A83675.1
 Restricted Item
Add to Cart
Add to Cart
AST-1306 (CAS 897383-62-9) is an inhibitor of epidermal growth factor receptor (EGFR) and HER2 receptor tyrosine kinases In biochemical assays AST-1306 inhibits EGFR and HER2 kinase activity with IC50 values of 0 5 nM and 3 nM respectively It also demonstrates inhibitory activity against EGFR mutations such as T790M/L858R reducing receptor phosphorylation and downstream signaling in both cell-free and cellular systems In various animal tumor models including ErbB2-overexpressing and SK-OV-3 xenografts AST-1306 suppresses tumor growth and signaling pathways supporting its use in EGFR- and HER2-related oncology research