Learn More
Selleck Chemical LLC Allitinib (AST-1306) 5mg 1050500-29-2 AST-6
SDP
Supplier:  Selleck Chemical LLC S21855MG
Encompass_Preferred
Allitinib (AST-1306, AST-6) is a novel irreversible inhibitor of EGFR and ErbB2 with IC50 of 0.5 nM and 3 nM, also effective in mutation EGFR T790M/L858R, more potent to ErbB2-overexpressing cells, 3000-fold selective for ErbB family than other kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2144