Learn More
Apexbio Technology LLC AST-1306, 5mg. Cas: 897383-62-9 MFCD: MFCD17169993. EGFR/HER2 inhibitor
SDP
Supplier:  Apexbio Technology LLC A83675
Encompass_Preferred
AST1306 is a novel inhibitor of EGFR and HER2 (IC50 = 0.5 nM and 3 nM respectively)EGFR (epidermal growth factor receptor) is a cell-surface receptor tyrosine kinase. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5129