Learn More
TARGETMOL CHEMICALS INC AST 487 10MG
SDP
Catalog No. 502576819
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AST 487 (NVP-AST 487) is a RET kinase inhibitor inhibiting RET autophosphorylation and activation of downstream effectors. It also can inhibit Flt-3. purity: 99%
Catalog No. 50-257-6819 Supplier TARGETMOL CHEMICALS INC Supplier No. T405310MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AST 487 (NVP-AST 487) is a RET kinase inhibitor inhibiting RET autophosphorylation and activation of downstream effectors. It also can inhibit Flt-3. purity: 99%