Learn More
Apexbio Technology LLC AST 487 630124-46-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092981
Encompass_Preferred
AST487 (CAS 630124-46-8) is a small molecule inhibitor primarily targeting RET kinase exhibiting an IC50 of 0 88 M As a member of the N N -diphenylurea class AST487 inhibits RET autophosphorylation and attenuates downstream signaling pathways such as PLC and ERK in a dose-dependent manner It also displays inhibitory activity against additional kinases including KDR Flt-3 and c-Kit In cellular models harboring RET activating mutations (e g C634W) AST487 suppresses RET-mediated signaling and reduces proliferation of thyroid carcinoma cells with RET mutations In xenograft mouse models oral administration of AST487 leads to a notable reduction in tumor volume making it valuable for preclinical studies of RET-driven cancers
Catalog No. 50-000-92981 Supplier Apexbio Technology LLC Supplier No. A31945.1
AST487 (CAS 630124-46-8) is a small molecule inhibitor primarily targeting RET kinase exhibiting an IC50 of 0 88 M As a member of the N N -diphenylurea class AST487 inhibits RET autophosphorylation and attenuates downstream signaling pathways such as PLC and ERK in a dose-dependent manner It also displays inhibitory activity against additional kinases including KDR Flt-3 and c-Kit In cellular models harboring RET activating mutations (e g C634W) AST487 suppresses RET-mediated signaling and reduces proliferation of thyroid carcinoma cells with RET mutations In xenograft mouse models oral administration of AST487 leads to a notable reduction in tumor volume making it valuable for preclinical studies of RET-driven cancers