Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AST 487 | 630124-46-8 | 98.90% | 529.56 | 1 ML
SDP

Catalog No. 5000360161
Encompass_Preferred

AST 487 is a RET kinase inhibitor that inhibits RET autophosphorylation and activation of downstream effectors. It also inhibits Flt-3. This compound is suitable for laboratory research due to its diverse inhibitory activities.

  • Inhibits RET (IC50=880 nM), KDR (IC50=170 nM), Flt-4 (IC50=790 nM), Flt-3 (IC50=520 nM), c-Kit (IC50=500 nM), and Abl (IC50=20 nM).
  • Potently inhibits the growth of human thyroid cancer cell lines with activating mutations of RET.
  • Markedly inhibits GDNF/GFRα1 and persephin-induced calcitonin mRNA at 100 nM in MTC-M cells.
  • Acts as a novel, mutant FLT3 inhibitor with a Ki of 0.12 μM.
  • Potently inhibits cellular proliferation (IC50<5 nM) in FLT3-ITD-Ba/F3 cells and D835Y-Ba/F3 cells.
  • Results in complete cell killing of FLT3-ITD-Ba/F3 cells at 0.01 μM.

Catalog No. 50-003-60161 Supplier Medchemexpress LLC Supplier No. HY1500210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AST 487 is a RET kinase inhibitor that inhibits RET autophosphorylation and activation of downstream effectors. It also inhibits Flt-3. This compound is suitable for laboratory research due to its diverse inhibitory activities.

  • Inhibits RET (IC50=880 nM), KDR (IC50=170 nM), Flt-4 (IC50=790 nM), Flt-3 (IC50=520 nM), c-Kit (IC50=500 nM), and Abl (IC50=20 nM).
  • Potently inhibits the growth of human thyroid cancer cell lines with activating mutations of RET.
  • Markedly inhibits GDNF/GFRα1 and persephin-induced calcitonin mRNA at 100 nM in MTC-M cells.
  • Acts as a novel, mutant FLT3 inhibitor with a Ki of 0.12 μM.
  • Potently inhibits cellular proliferation (IC50<5 nM) in FLT3-ITD-Ba/F3 cells and D835Y-Ba/F3 cells.
  • Results in complete cell killing of FLT3-ITD-Ba/F3 cells at 0.01 μM.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.