Learn More
Selleck Chemical LLC AST-487
SDP
Supplier:  Selleck Chemical LLC S66625MG
Encompass_Preferred
AST-487 (NVP-AST487) a N N -diphenyl urea is an ATP competitive inhibitor of Flt3 with ki of 0 12 M Besides FLT3 AST487 also inhibits RET KDR c-KIT and c-ABL kinase with IC50 values below 1 M
Catalog No. 50-000-02372