Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics AST-GN QC Set, 7 Strains
SDP

Catalog No. 22025863 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-025-863 2 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-025-863 Supplier Microbiologics Supplier No. 5260P
Add to Cart
Add to Cart

This QC set is designed for testing applications involving seven strains.

AST-GN QC Set, 7 strains contains two KWIK-STIKs of each strain listed below (14 KWIK-STIKs total).

  • 0335P Escherichia coli derived from ATCC™ 25922™
  • 0495P Escherichia coli derived from ATCC™ 35218™
  • 01005P Klebsiella pneumoniae derived from ATCC™ BAA-1705™
  • 01263P Klebsiella pneumoniae derived from ATCC™ BAA-2814™
  • 0784P Klebsiella quasipneumoniae subsp. similipneumoniae derived from ATCC™ 700603™
  • 0353P Pseudomonas aeruginosa derived from ATCC™ 27853™
  • 01290P Pseudomonas aeruginosa derived from ATCC™ BAA-3144™

Specifications

Form Lyophilized
Manufacturing Origin United States
Product Type Bacterial Growth Medium
Sterility Non-sterile
Target Organism Class Anaerobes
Test Type Antimicrobial Susceptibility, Antimicrobial Susceptibility for Gram-positive Organisms, Microbial Identification, Qualitative QC
Quantity 2 Pk.
Species Escherichia coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.