Learn More
Teco Diagnostics AST/GOT LIQUID(KINETIC)150TEST
Catalog No. NC9851324
Encompass_Preferred
For the quantitative determination of aspartate aminotransferase (AST) in human serum. The reagents are used in routine examination and monitoring of therapy and relapses. 1x125mL Bottle of ALT(SGOT), Liquid Reagent 1 1x25mL Bottle of ALT(SGOT), Liquid Reagent 2
Catalog No. NC9851324 Supplier Teco Diagnostics Supplier No. A559150
For the quantitative determination of aspartate aminotransferase (AST) in human serum. The reagents are used in routine examination and monitoring of therapy and relapses. 1x125mL Bottle of ALT(SGOT), Liquid Reagent 1 1x25mL Bottle of ALT(SGOT), Liquid Reagent 2