Learn More
Medchemexpress LLC AST5902 trimesylate | 2929417-90-1 | 98.75% | 842.88 | 100 MG
SDP
Catalog No. 5000352462
Encompass_Preferred
AST5902 trimesylate is the principal metabolite of Alflutinib in vitro and in vivo. It exhibits antineoplastic activity, and Alflutinib is known as an EGFR inhibitor.
  • Principal metabolite of Alflutinib
  • Exhibits antineoplastic activity
  • Targets EGFR
Catalog No. 50-003-52462 Supplier Medchemexpress LLC Supplier No. HY138627A100MG
AST5902 trimesylate is the principal metabolite of Alflutinib in vitro and in vivo. It exhibits antineoplastic activity, and Alflutinib is known as an EGFR inhibitor.
  • Principal metabolite of Alflutinib
  • Exhibits antineoplastic activity
  • Targets EGFR