Learn More
Medchemexpress LLC AST5902 trimesylate | 2929417-90-1 | 98.8% | 842.88 | 50 MG
SDP
Catalog No. 5000350435
Encompass_Preferred
AST5902 trimesylate is the principal metabolite of Alflutinib (AST2818) both in vitro and in vivo. It exerts antineoplastic activity. Alflutinib is an EGFR inhibitor.
  • Principal metabolite of Alflutinib
  • Exerts antineoplastic activity
  • EGFR inhibitor
  • Yellow to orange solid
  • Store at 4°C, sealed, away from moisture and light
  • In solvent: -80°C for 6 months; -20°C for 1 month
Catalog No. 50-003-50435 Supplier Medchemexpress LLC Supplier No. HY138627A50MG
AST5902 trimesylate is the principal metabolite of Alflutinib (AST2818) both in vitro and in vivo. It exerts antineoplastic activity. Alflutinib is an EGFR inhibitor.
  • Principal metabolite of Alflutinib
  • Exerts antineoplastic activity
  • EGFR inhibitor
  • Yellow to orange solid
  • Store at 4°C, sealed, away from moisture and light
  • In solvent: -80°C for 6 months; -20°C for 1 month