Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AST5902 trimesylate | 2929417-90-1 | 98.8% | 842.88 g/mol | C30H41F3N8O11S3 | 5 MG
SDP

Catalog No. 5000221235
Encompass_Preferred

AST5902 trimesylate is the primary metabolite of the EGFR inhibitor alflutinib and exhibits antineoplastic activity. Supplied as a research standard (CAS 2929417-90-1), it is intended for in vitro and in vivo research use such as metabolite profiling, pharmacology, and analytical reference studies.

  • High purity suitable for analytical and biological research.
  • Molecular weight 842.88 g/mol and formula C30H41F3N8O11S3.
  • Available as a solid and as ready-to-use DMSO solutions.
  • Intended for in vitro and in vivo research applications only.
  • Appropriate as a reference standard for metabolite and pharmacokinetic studies.

Catalog No. 50-002-21235 Supplier Medchemexpress LLC Supplier No. HY138627A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AST5902 trimesylate is the primary metabolite of the EGFR inhibitor alflutinib and exhibits antineoplastic activity. Supplied as a research standard (CAS 2929417-90-1), it is intended for in vitro and in vivo research use such as metabolite profiling, pharmacology, and analytical reference studies.

  • High purity suitable for analytical and biological research.
  • Molecular weight 842.88 g/mol and formula C30H41F3N8O11S3.
  • Available as a solid and as ready-to-use DMSO solutions.
  • Intended for in vitro and in vivo research applications only.
  • Appropriate as a reference standard for metabolite and pharmacokinetic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.