Learn More
Medchemexpress LLC aStAx-35R TFA | 99.1% | 2392.86 | 1 MG
SDP
Catalog No. 5000425113
Encompass_Preferred
aStAx-35R TFA is a stapled peptide that functions as an antagonist of the nuclear form of β-catenin, thereby inhibiting Wnt signaling. It competitively prevents the binding of β-catenin to TCF4 and selectively induces growth inhibition in Wnt-dependent cancer cells.
  • Directly targets and antagonizes the nuclear form of β-catenin.
  • Effectively inhibits the Wnt signaling pathway.
  • Competitively inhibits the binding of β-catenin to TCF4.
  • Selectively induces growth inhibition in cancer cells dependent on Wnt signaling.
Catalog No. 50-004-25113 Supplier Medchemexpress LLC Supplier No. HYP10392B1MG
aStAx-35R TFA is a stapled peptide that functions as an antagonist of the nuclear form of β-catenin, thereby inhibiting Wnt signaling. It competitively prevents the binding of β-catenin to TCF4 and selectively induces growth inhibition in Wnt-dependent cancer cells.
  • Directly targets and antagonizes the nuclear form of β-catenin.
  • Effectively inhibits the Wnt signaling pathway.
  • Competitively inhibits the binding of β-catenin to TCF4.
  • Selectively induces growth inhibition in cancer cells dependent on Wnt signaling.