Learn More
TARGETMOL CHEMICALS INC ASTAXANTHIN 50MG
SDP
Catalog No. 502576994
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Astaxanthin (trans-Astaxanthin) is a keto-carotenoid in the terpenes class of chemical compounds. It is classified as a xanthophyll but it is a carotenoid with no vitamin A activity. It is present in most red-colored aquatic organisms. Astaxanthin has shown to mediate anti-oxidant and anti-inflammatory actions. It may be found in fish feed or some animal food as a color additive. purity: 99%
Catalog No. 50-257-6994 Supplier TARGETMOL CHEMICALS INC Supplier No. T313850MG
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Astaxanthin (trans-Astaxanthin) is a keto-carotenoid in the terpenes class of chemical compounds. It is classified as a xanthophyll but it is a carotenoid with no vitamin A activity. It is present in most red-colored aquatic organisms. Astaxanthin has shown to mediate anti-oxidant and anti-inflammatory actions. It may be found in fish feed or some animal food as a color additive. purity: 99%