Learn More
AdipoGen Astaxanthin (5 mg)
SDP
Supplier:  AdipoGen AGCN20055M005
Encompass_Preferred
Chemical. CAS: 472-61-7. Formula: C40H52O4. MW: 596.8. Synthetic. Potent antioxidant carotenoid. Free radical scavenger. Reduces oxidative stress. Anticancer compound. Anti-inflammatory. Blocks NF-kappaB activation. Anti-hypertensive. Cardiovascular protective. Improves neuronal stem cell potential. Neuroprotective. Inhibits tumor invasion. Apoptosis inducer . Increases insulin sensitivity. Attenuates diabetes associated coagulatory, oxidative and inflammatory stress. PPARalpha agonist and PPARgamma antagonist. Shows positive effects on obesity and insulin resistance Hepatoprotective. Reviews.
Catalog No. 50-205-3400