Learn More
Medchemexpress LLC Astaxanthin (rac./meso) | 7542-45-2 | 596.84 g/mol | C40H52O4 | 100mg
SDP
Catalog No. 5000268291
Encompass_Preferred
(rac /meso)-Astaxanthin (Standard) is the analytical standard of (rac /meso)-Astaxanthin This product is intended for research and analytical applications rac-Astaxanthin is a carotenoid pigment found primarily in marine animals including shrimp and salmon It is a potent lipid-soluble antioxidant
Catalog No. 50-002-68291 Supplier Medchemexpress LLC Supplier No. HYW750676R100MG
(rac /meso)-Astaxanthin (Standard) is the analytical standard of (rac /meso)-Astaxanthin This product is intended for research and analytical applications rac-Astaxanthin is a carotenoid pigment found primarily in marine animals including shrimp and salmon It is a potent lipid-soluble antioxidant