Learn More
Aviva Systems Biology Corp Aste1 Antibody - middle region (ARP37917_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37917_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Aste1. It was validated on Western Blot. Aste1,asteroid homolog 1 (Drosophila),AV006403; MGC107393; 1100001A21Rik; Aste1,protein asteroid homolog 1,protein asteroid homolog 1,OTTMUSP00000025869The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1100001A21Rik, AV006403, MGC107393ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-284-702