Learn More
Medchemexpress LLC Astegolimab, 5mg
SDP
Supplier:  Medchemexpress LLC HYP994445MG
Encompass_Preferred
Astegolimab (MSTT 1041A; RG 6149) is a human IgG2 monoclonal antibody that blocks IL-33 signaling by targeting ST2, the IL-33 receptor. Astegolimab has the potential for chronic obstructive pulmonary disease (COPD) research.
Catalog No. NC2810473