Learn More
Medchemexpress LLC HY-12532 5mg Medchemexpress, Astemizole CAS:68844-77-9 Purity:>98%
SDP
Catalog No. 501932603
Encompass_Preferred
Medchemexpress, HY-12532 5mg Astemizole CAS:68844-77-9 Astemizole (R 43512), a second-generation antihistamine drug to diminish allergic symptoms with a long duration of action, is a histamine H1-receptor antagonist, with an IC 50 of 4 nM. Astemizole also shows potent hERG K + channel blocking activity with an IC 50 of 0.9 nM. Astemizole has antipruritic effects [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2603 Supplier Medchemexpress LLC Supplier No. HY125325MG
Medchemexpress, HY-12532 5mg Astemizole CAS:68844-77-9 Astemizole (R 43512), a second-generation antihistamine drug to diminish allergic symptoms with a long duration of action, is a histamine H1-receptor antagonist, with an IC 50 of 4 nM. Astemizole also shows potent hERG K + channel blocking activity with an IC 50 of 0.9 nM. Astemizole has antipruritic effects [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.