Learn More
Abcam Astemizole, histamine H1 receptor antagonist and hERG K+ channel blocker, 50MG
Catalog No. 502705600
Encompass_Preferred
MW 458.6 Da, Purity >99%. Potent, selective histamine H1 receptor antagonist (IC50 = 4 nM) and potent hERG K+ channel blocker (IC50 =0.9 nM). Orally active. Antiallergic agent. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5600 Supplier Abcam Supplier No. AB14458350MG
MW 458.6 Da, Purity >99%. Potent, selective histamine H1 receptor antagonist (IC50 = 4 nM) and potent hERG K+ channel blocker (IC50 =0.9 nM). Orally active. Antiallergic agent. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement