Learn More
Medchemexpress LLC HY-N0509 10mg Medchemexpress, Astilbin CAS:29838-67-3 Purity:>98%
SDP
Catalog No. 501933598
Encompass_Preferred
Medchemexpress, HY-N0509 10mg Astilbin CAS:29838-67-3 Astilbin, a flavonoid compound, is isolated from the rhizome of Smilax glabra. Astilbin enhances NRF2 activation. Astilbin also suppresses TNF-α expression and NF-κB activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3598 Supplier Medchemexpress LLC Supplier No. HYN050910MG
Medchemexpress, HY-N0509 10mg Astilbin CAS:29838-67-3 Astilbin, a flavonoid compound, is isolated from the rhizome of Smilax glabra. Astilbin enhances NRF2 activation. Astilbin also suppresses TNF-α expression and NF-κB activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.