Learn More
Cayman Chemical Astlbn 10mg
SDP
Catalog No. 502940757
Encompass_Preferred
An inhibitor of plasma kallikrein (Ki 0 81 M) reduces bradykinin generation induced by kaolin and -carrageenan ex vivo in human plasma prolongs partial thromboplastin and euglobulin clot lysis times reduces hyperplasia pannus formation and infiltration of inflammatory cells in the tarsal joint of mice with collagen-induced arthritis
Catalog No. 50-294-0757 Supplier Cayman Chemical Supplier No. 28267 10MG
An inhibitor of plasma kallikrein (Ki 0 81 M) reduces bradykinin generation induced by kaolin and -carrageenan ex vivo in human plasma prolongs partial thromboplastin and euglobulin clot lysis times reduces hyperplasia pannus formation and infiltration of inflammatory cells in the tarsal joint of mice with collagen-induced arthritis