Learn More
Thermco Products Inc ASTM 120C, Kenimatic Viscosity, 38.6/41.4C, 0.05C div, TOTAL imm, 310mm, certified at 0 deg.C, 40 deg.C, 41 deg.C
Catalog No. NC0670098
Encompass_Preferred
ASTM 120C thermometer certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.
Catalog No. NC0670098 Supplier Thermco Products Inc Supplier No. ACC120CFC
ASTM 120C thermometer certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.