Learn More
Supply Solutions Thermco - 400Mm Mercury Thermometer Astm. Immersion Depth: Total; Overall Length: 400Mm; Temp Range: 30 To 180F
Catalog No. NC0889148
Encompass_Preferred
These liquid filled glass thermometers are manufactured to very rigid specifications and are guaranteed to meet accuracy requirements set forth by ASTM. The suffix portion of the catalog number is the ASTM designation for that particular thermometer. Factory calibration certificates are available for an additional charge. Please specify calibration points at time of order.; Immersion Depth: Total; Overall Length: 400mm; Temp Range: 30 to 180F; Length (MM): 397; Immersion (MM): Total
Catalog No. NC0889148 Supplier Supply Solutions Supplier No. TMC ACC015F
These liquid filled glass thermometers are manufactured to very rigid specifications and are guaranteed to meet accuracy requirements set forth by ASTM. The suffix portion of the catalog number is the ASTM designation for that particular thermometer. Factory calibration certificates are available for an additional charge. Please specify calibration points at time of order.; Immersion Depth: Total; Overall Length: 400mm; Temp Range: 30 to 180F; Length (MM): 397; Immersion (MM): Total