Learn More
ASTM™ 2-Bromoethanol Solution, 2,000 μg/mL in Toluene, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| Product Line | ASTM |
| CAS | 540-51-2 |
| Flash Point | 4°C |
| Linear Formula | BrCH2 CH2 OH |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Formula Weight | 124.96 |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 2,000 μg/mL in toluene |
ASTM is a registered trademark of American Society for Testing and Materials
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.