Learn More
Crescent Chemical Co Inc ASTM 3710 SIM DISTIL 16COMP1ML
SDP
Catalog No. 5068259
Encompass_Preferred
ASTM Method D3710 - Simulated Distillation Calibration Mix, 16 Components n-Butylbenzene-3.5%, n-Decane-3.5%, 2,4-Dimethylpentane-5.8%, n-Dodecane-3.5%, n-Heptane-10.5%, n-Hexane-5.8%, 2-Methylbutane-10.5%, 2-Methylpentane-5.8%, n-Octane-5.8%, n-Pent
Catalog No. 50-682-59 Supplier Crescent Chemical Co Inc Supplier No. CTCC2374
ASTM Method D3710 - Simulated Distillation Calibration Mix, 16 Components n-Butylbenzene-3.5%, n-Decane-3.5%, 2,4-Dimethylpentane-5.8%, n-Dodecane-3.5%, n-Heptane-10.5%, n-Hexane-5.8%, 2-Methylbutane-10.5%, 2-Methylpentane-5.8%, n-Octane-5.8%, n-Pent