Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASTM™ 5442 (C12-C60) Quantitative Linearity Standard, 0.01% (w/w) Each Component in Cyclohexane, MilliporeSigma™ Supelco™

Catalog No. 111016424 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
6 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6424 6 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6424 Supplier MilliporeSigma Supelco Supplier No. 502235
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Concentration 0.01% (w/w) each component in cyclohexane
Quantity 6 x 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Docosane; Dodecane; Dotriacontane; Eicosane; Hexacontane; Hexacosane; Hexadecane; Hexatriacontane; Octacosane; Octadecane; Pentacontane; Tetracontane; Tetracosane; Tetradecane; Tetratetracontane; Triacontane

ASTM is a registered trademark of American Society for Testing and Materials

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.