Learn More
Troemner™ ASTM Analytical Precision Class 2 Weight Set with ISO/IEC 17025 Accredited Certificate
Catalog No. 02218295 Shop All Troemner Products
Change view
Click to view available options
Weight Set; Weight: 10 mg to 100 g; Material: Troemner Alloy 8, Stainless Steel
Catalog No.
02-218-295 Weight Set; Weight: 10 mg to 100 g; Material: Troemner Alloy 8, Stainless Steel
1 options
Catalog No. 02-218-295 Supplier Troemner™ Supplier No. 80859288
Add to Cart
Add to Cart
Troemner Calibration Weight Sets Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Certifications/Compliance ISO/IEC 17025 Accredited Certificate
Material Troemner Alloy 8, Stainless Steel
Weight (Metric) 10 mg to 100 g
Shape Weights 50 kg Through 1 g are Cylindrical with Handling Knob; Weights 500 mg and Below are Flat with One End Turned Up for Easy Handling
Includes (2) 2 g; (2) 20 mg; 500 mg; 10 g; 100 g; (2) 20 g; 100 mg; 10 mg; (2) 200 mg; 1 g; 50 g; 5 g; 50 mg