Learn More
Troemner™ ASTM Class 2 Weight with Traceable Certificate
Catalog No. 02218224 Shop All Troemner Products
Change view
Click to view available options
Calibration Weight; Weight: 2 g
Catalog No.
02-218-224 Calibration Weight; Weight: 2 g
1 options
Catalog No. 02-218-224 Supplier Troemner™ Supplier No. 80858287
Add to Cart
Add to Cart
Troemner Calibration Weight Sets Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Certifications/Compliance Traceable Certificate
Material Troemner Alloy 8
Case Material Polycarbonate
Weight (Metric) 2 g
Shape Cylindrical