Learn More
Troemner™ ASTM Class 4 Weight Set with ISO/IEC 17025 Accredited Certificate
Catalog No. 02218314 Shop All Troemner Products
Change view
Click to view available options
Weight Set; Weight: 1 to 500 mg
Catalog No.
02-218-314 Weight Set; Weight: 1 to 500 mg
1 options
Catalog No. 02-218-314 Supplier Troemner™ Supplier No. 80850263
Add to Cart
Add to Cart
Troemner Calibration Weight Sets Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Certifications/Compliance ISO/IEC 17025 Accredited Certificate
Material Stainless Steel, Aluminum
Case Material Polycarbonate
Weight (Metric) 1 to 500 mg
Density Below 5 mg are 2.70 g/cm3; 500 to 5 mg are 7.95 g/cm3
Height (English) Case 4.5 in.
Height (Metric) Case 11.43 cm
Includes 100 mg; 10 mg; (2) 200 mg; (2) 20 mg; 5 mg; (2) 2 mg; 50 mg; 500 mg; 1 mg