Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASTM™ D2887 Calibration Mix Analytical Standard, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022585 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
6 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2585 6 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2585 Supplier MilliporeSigma Supelco Supplier No. 48882
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aroclor 1248 is a light yellow to yellow-green colored, commercially available polychlorinated biphenyl product liquid mixture. It consists of tri-, tetra-, and pentachloro congeners.

TRUSTED_SUSTAINABILITY

Specifications

Flash Point -22°C (Closed Cup, Not Applicable)
UN Number UN3295 - Class 3 - PG 2 - EHS - VP > 110 kPa - Hydrocarbons, liquid, n.o.s., HI: A, S (not BR)
Quantity 6 x 1 mL
Grade Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.