Learn More
ASTM™ D3710 Qualitative Calibration Mix, Analytical Standard, MilliporeSigma™ Supelco™
Catalog No. 111016427 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
6 x 1 mL
Catalog No. Quantity
11-101-6427 6 x 1 mL
1 options
Catalog No. 11-101-6427 Supplier MilliporeSigma Supelco Supplier No. 48884
Specifications
Quantity 6 x 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Butane % (w/w); Butylbenzene 3.2 wt. %; Decane 3.2 % (w/w); 2,4-Dimethylpentane 5.4 % (w/w); Dodecane 3.2 % (w/w); Heptane 9.7 % (w/w); Hexane 5.4 % (w/w); 2-Methylbutane 9.7 % (w/w); 2-Methylpentane 5.4 % (w/w); 2-Methylpropane 1.5 % (w/w); Octane 5.4 % (w/w); Pentadecane 2.2 % (w/w); Pentane 7.6 % (w/w); Propane 1.5 % (w/w); Propylbenzene 4.3 % (w/w); Tetradecane 2.2 % (w/w); Toluene 10.8 % (w/w); Tridecane 2.2 % (w/w); p-Xylene 13 % (w/w)
ASTM is a registered trademark of American Society for Testing and Materials