Learn More
METASPEC ASTM D4340 Heat-Transfer Cast Aluminum Test Specimen 10PK. Each specimen has two drilled edge holes for thermocouples, and one face polished per ASTM D4340. Sold in multiples of 10.
Catalog No. NC3959119
Encompass
Cast aluminum test specimen (also known as a heat-transfer specimen) used in ASTM D4340 (Corrosion of Cast Aluminum Alloys in Engine Coolants Under Heat-Rejecting Conditions). The metal test specimen is 2.6" in diameter and 1/2" thick and has two thermocouple holes drilled into one edge. The material is 319 cast aluminum (UNS A03190). ASTM D4340 is run in duplicate; two test specimens are required.
Customized test specimens for modified versions of this test method are available on request.
Catalog No. NC3959119 Supplier METASPEC Supplier No. 3024340319
Cast aluminum test specimen (also known as a heat-transfer specimen) used in ASTM D4340 (Corrosion of Cast Aluminum Alloys in Engine Coolants Under Heat-Rejecting Conditions). The metal test specimen is 2.6" in diameter and 1/2" thick and has two thermocouple holes drilled into one edge. The material is 319 cast aluminum (UNS A03190). ASTM D4340 is run in duplicate; two test specimens are required.
Customized test specimens for modified versions of this test method are available on request.