Learn More
ASTM™ D5134 Splitter Linearity Check Mix Analytical Standard, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Flash Point | -11.1°C (Closed Cup, Not Applicable) |
| UN Number | UN3295 - Class 3 - PG 2 - EHS - VP > 110 kPa - Hydrocarbons, liquid, n.o.s., HI: A, S (not BR) |
| Quantity | 1 mL |
| Grade | Analytical Standard |
ASTM is a registered trademark of American Society for Testing and Materials
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.