Learn More
ASTM™ D5307 Crude Oil Internal Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111004630 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mL
Catalog No. Quantity
11-100-4630 25 mL
1 options
Catalog No. 11-100-4630 Supplier MilliporeSigma Supelco Supplier No. 48479
Add to Cart
Add to Cart
Equal weights of the hydrocarbons listed. Add to the crude oil sample as the internal standard.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 25 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Heptadecane 25 % (w/w); Hexadecane 25 % (w/w); Pentadecane 25 % (w/w); Tetradecane 25 % (w/w)
ASTM is a registered trademark of American Society for Testing and Materials