Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASTM™ D5442 C16-C44 Quantitative Linearity Analytical Standard, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022611 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
6 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2611 6 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2611 Supplier MilliporeSigma Supelco Supplier No. 502286
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Flash Point -18°C (Closed Cup)
UN Number UN1145 - Class 3 - PG 2 - Cyclohexane
Quantity 6 x 1 mL
Grade Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.