Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASTM™ D6584 1,2,4-Butanetriol Solution, 1,000 μg/mL in Pyridine, MilliporeSigma™ Supelco™

Catalog No. 111017770 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7770 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7770 Supplier MilliporeSigma Supelco Supplier No. CRM44896
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 42890-76-6
Product Line ASTM
Flash Point 17.0°C - closed cup
Concentration 1000 μg/mL in pyridine
Synonym Biodiesel impurity; Biodiesel impurity; Butanetriol solution
Type Certified Reference Material
Quantity 5 mL
Grade TraceCERT™ Analytical Standard

ASTM is a registered trademark of American Society for Testing and Materials

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.