Learn More
ASTM™ D6584 Standard Solution 5, Certified Reference Material, in Pyridine (varied), MilliporeSigma™ Supelco™
Catalog No. 111016444 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
11-101-6444 1 mL
1 options
Catalog No. 11-101-6444 Supplier MilliporeSigma Supelco Supplier No. CRM44917
Add to Cart
Add to Cart
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 1,3-Diolein 500 μg/mL; Glycerol 50 μg/mL; Monolein 1000 μg/mL; Triolein 500 μg/mL
ASTM is a registered trademark of American Society for Testing and Materials